N-Tetradecylphosphocholine ≥99.5%, Crystallization grade

Supplier: CUBE BIOTECH

Synonyms: Tetradecylphosphocholine, Fos-Choline™ 14, Fos-14

16029 16030 16031
77392-356PK 235.8 USD
77392-356 77392-358 77392-360
N-Tetradecylphosphocholine ≥99.5%, Crystallization grade
N-Tetradecylphosphocholine

Phosphocholines, or Fos-cholines, are a novel class of detergents that is well suited for the solubilization, stabilization, and purification of membrane proteins. For screening of the best-suited detergent for a given application, we offer Fos-cholines with chain lengths from C-9 to C-16.


Molecular Weight: 379.5 g/mol
Form: White crystalline powder
CMC (in H2O): 0.12 mM (0.0046%)
Synonyms are: Tetradecylphosphocholine, Fos-14

MW: 379.5 g/mol
CAS Number: 82494-09-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37173ada861915e3c019a858c07e19b93dc44f62&vid=75ac6e37e8c3947b11538986228396af76c860c1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR