Stilla® Sapphire Chip
Supplier: STILLA TECHNOLOGIES, INC.
C14012
77392-264PK 435.76 USD
77392-264
Stilla® Sapphire Chip
Nucleic Acid Reagents End-point PCR Enzymes and Kits
The Sapphire Chip features cutting-edge, patented microfluidic technology that integrates the entire digital PCR workflow into a single consumable.
  • Capacity: Up to four samples/chip
  • Input volume: 25 µl/chamber
  • Droplets per sample: Up to 30000
  • Dynamic range: ∼5 log
For maximum template input volume, the Sapphire Chip can be loaded with up to 4 samples per chip, and generate up to 30,000 droplets per sample for your digital PCR workflows. After sample preparation, just a single pipetting step is required before the chip is loaded onto either the 3-color naica® system or the 6-color naica® system for partitioning, PCR amplification, and sample read-out. The Sapphire Chip supports applications such as rare mutation detection, liquid biopsy analysis, gene expression analysis, and pathogen detection.