DIGE LF24 Precast Gels
Supplier: Cytiva
29706670
77392-084EA 1253.98 USD
77392-084
DIGE LF24 Precast Gels
Electrophoresis Gels Precast Gels
DIGE Gels LF24 are large format precast 12.5% polyacrylamide gels with low fluorescent glass plates intended for 2D DIGE and 2D DIBE™.
  • Ready to use precast gel
  • Quick set up with optimized DIGE buffer kit
  • Optimized 24 cm IPG strips for large format protein separation
  • Low fluorescent glass plates enable direct fluorescent image scan
  • Suitable for large format HCP coverage assay for regulatory requirement of HCP ELISA
  • Gel cassette: Low fluorescent glass
DIGE Gels LF24 are large format precast 12.5% polyacrylamide gels with low fluorescent glass plates intended for 2D DIGE/DIBE™ with Amersham™ DIGE Unit LF24 electrophoresis.
The glass plates do not need to be removed when scanning the gel with a fluorescent image scanner. Open the glass cassette to perform transfer of the proteins to a membrane.
This gel is intended for two-dimensional fluorescence difference gel electrophoresis (2-D DIGE) and large format HCP coverage assay (2D DIBE™) for regulatory requirements of HCP ELISA. Ready to use DIGE buffer kits allow for a quick electrophoresis set up. The package contains three precast gels.