Linearity FLQ HbA1c for Bio-Rad Systems
Supplier: AUDIT MICROCONTROLS
K761M-5
76545-590EA 459.19 USD
76545-590 77391-858
Linearity FLQ HbA1c for Bio-Rad Systems
Controls and Calibration Verification Materials
Linearity FLQ HbA1c for Bio-Rad® Systems is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for HbA1c.
  • Linearity; Frozen Liquid
The Audit® MicroControlsTM Linearity FLQ HbA1c is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analyte: HbA1c.
Delivery information: Priority overnight.
Packaging: Kits are shrinked-wrapped.