4-Bromo-2-methyl-2'-(trifluoromethyl)-1,1'-biphenyl ≥97%
Supplier: AOBChem USA
|
Warning
|
56863-500MG
56863-1G
56863-5G
56863-25G
77329-770EA
184.37
USD
77329-770
77347-162
77366-050
77387-868
4-Bromo-2-methyl-2'-(trifluoromethyl)-1,1'-biphenyl ≥97%
4-Bromo-2-methyl-2'-(trifluoromethyl)-1,1'-biphenyl
Caution: for Research Use Only
Formula:
C₁₄H₁₀BrF₃ Storage Temperature: Ambient |
MDL Number:
MFCD34530550 CAS Number: 2734773-86-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...