1-Bromo-2-chloro-4-ethoxy-5-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

60572-500MG 60572-1G 60572-5G
77340-320EA 900.22 USD
77340-320 77355-158 77379-246
1-Bromo-2-chloro-4-ethoxy-5-methoxybenzene ≥97%
1-Bromo-2-chloro-4-ethoxy-5-methoxybenzene

Caution: for Research Use Only

Formula: C₉H₁₀BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD34759830
CAS Number: 2386542-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR