PINK UNDERGUARD* Nitrile Examination Gloves, Powder-free, Latex-free
Supplier: O&M HALYARD, INC
47452 47453 47454 47455 47456
77322-590BX 65.75 USD
77322-590 77322-592 77322-594 77322-596 77322-598
PINK UNDERGUARD* Nitrile Examination Gloves, Powder-free, Latex-free
Gloves
The first medical examination glove specifically designed to help healthcare professionals comply with recommended double gloving protocols when handling chemotherapy drugs.
  • Coated exterior of the glove reduces friction
  • Coating makes donning a second pair of gloves easy and alleviates the typical frustrations associated with double glovings
  • Tested with 52 chemotherapy drugs in accordance with ASTM D6978
  • Extended 12" cuff, providing additional coverage under the cuff of the chemo gown
  • Pink color provides visible breach detection when wearing contrasting outer gloves
As new PPE protocols for handling chemotherapy agents are implemented, it is imperative to have a chemotherapy glove that appropriately protects healthcare professionals and makes compliance to double gloving easy. Powder-free PINK UNDERGUARD* nitrile examination gloves do both.