Stainless Steel Wire Cage Lids, Tecniplast®
Supplier: Tecniplast Usa
1264C116 1354G144 2000P214 2000P214 1144B116 1284L116 1354G115 1354G117 2000P242 2000P962 2154F114 1264C116
10713-102CS 4548.2 USD
10713-102 10713-146 10713-270 10713-340 21020-308 21020-309 77322-366 77322-386 77322-400 77322-402 77322-410 77322-414 77322-426 77322-376
Stainless Steel Wire Cage Lids, Tecniplast®
Animal Supplies Animal Cages
Desgined to ensure a perfect fit and secure lock throughout the complete range of Tecniplast® cages.
Stainless steel drop-in hoppers are autoclavable. A central divider prevents the animal from crawling into the hoppper, which can be used with or without a raised wire floor.
Secure and easy to handle, these autoclavable stainless steel wire lids have cross-wire members to prevent bending. With the exception of flat lids 95057-874 and 95057-888, all lids feature a U-shaped hopper for easy access to food. Hoppers won’t entrap food or contaminants. Wire spacing is 7 mm for most sizes; 19 mm spacing for guinea pig cage lid 10713-340. Hopper divider folds down, allowing lids to be stacked for storage. Flat lids 95057-874, 95057-888, and 10713-345 are for use without a raised floor. Incorporate stopper guard for water bottle sipper tube.