Pipet Keeper™ XL Biohazard Waste Container
Supplier: Whitney Products
BH2016
77321-812CS 416.73 USD
77321-812
Pipet Keeper™ XL Biohazard Waste Container
Boxes Waste Disposal Boxes
Pipet Keeper XL biohazard collection containers are designed to conveniently collect up to 16" long biohazardous waste at your workstation.
  • Dispose of pipettes, swabs and other long, biohazardous objects easily
  • Sturdy construction - made of sturdy paperboard printed with orange biohazard symbols
  • Plastic-lined interior holds a moderate amount of liquid
  • Self-locking tops keep contents safely enclosed
  • Flat storage requires far less space than rigid plastic containers
  • Interlocking flaps - easily fold into shape
Collects waste up to 16" - allows quick and efficient disposal without the need for additional handling or transportation.