Avantor® ACE® PFAS Delay Columns
Supplier: Avantor
ACE-PFASD-0502
77321-796EA 731.06 USD
77321-796
Avantor® ACE® PFAS Delay Columns
Chromatography Columns
Background PFAS (Perfluoronated Alkyl Substances) from an LC instrument and mobile phase can interfere with the detection of PFAS in samples. The introduction of a delay column, which is plumbed before the autosampler in the LC system, will delay background PFAS to improve accuracy of PFAS measurement by LC-MS.​
  • Silica-based fully porous particle delay column
  • Universal, can be used in a workflow setup with any analytical column format/dimensions
Allows for the analysis of PFAS from the sample, without interfering PFAS compromising the analysis (present and introduced by the instrumentation). Moreover, the data output/chromatogram benefits with the respective PFAS sample peaks separated/delayed from the background PFAS peaks.