VWR® Laboratory Chest Freezer with Natural Refrigerants, 20 cu. ft.
Supplier: VWR International
77321-624
77321-624EA 3448.7 USD
77321-624
VWR® Laboratory Chest Freezer with Natural Refrigerants, 20 cu. ft.
Freezers Laboratory / General Purpose Freezers
This versatile chest freezer delivers scientific precision and superior temperature control to laboratory environments.
  • Microprocessor temperature control
  • Temperature and power failure alarms
  • High density foam insulation
  • Keyed door lock
  • Natural hydrocarbon refrigerants
Designed for research and preservation, this chest freezer offers a spacious 20 cubic feet of storage capacity to efficiently organize samples. Preserve the integrity and longevity of delicate materials with trusted microprocessor control. Enjoy an intuitive user experience, reinforced construction, and energy-efficient design for cost savings and sustainability. Additionally, the use of hydrocarbon refrigerants brings you energy savings and environmental protection. Safeguard your research while maintaining the highest storage standards with this trusted product.
Use/purpose: Laboratory/pharmaceutical