Corepoint® Scientific 16 CF Refrigerator and Freezer Combo Unit
Supplier: Horizon Scientific
PRF162WWW/0HC
77320-856EA 7551.41 USD
77320-856
Corepoint® Scientific 16 CF Refrigerator and Freezer Combo Unit
Refrigerators
These comprehensive cold storage units deliver next-level temperature control to laboratory environments.
  • Superior temperature uniformity
  • Microprocessor temperature controllers
  • Fully array of alarms
  • Forced draft air circulation
  • Natural hydrocarbon refrigerants
These refrigerator and freezer combination units are designed with variable speed compressors to deliver unparalleled temperature control. Safeguard your samples with superior temperature uniformity and recovery, even after frequent door openings. Monitor your laboratory equipment though intuitive digital displays and protective alarms. Champion sustainability with natural hydrocarbon refrigerants - which not only contribute to energy savings, but also foster a greener laboratory footprint. And enjoy the convenience of auto-defrosting, eliminating the hassle of manual maintenance. *Elevate your laboratory cooling experience with an integrated solution that brings together innovation, performance, and precision.
Certifications: ETL/cETL Listed.
Caution: *Disclaimer: Auto-defrost freezers incorporate an electric heater installed on the evaporator that assists in the removal of the frost and ice. Although the heater helps speed up the frost and ice removal, it can raise the temperatures within the freezer. These air temperature spikes can be harmful to some medical and laboratory products.