Epredia™ SlideMate™ Laser Slide Printer
Supplier: Erie Scientific
8000048
77320-770EA 35920.97 USD
77320-770
Epredia™ SlideMate™ Laser Slide Printer
Printers Cassette and Slide Printers
Enter a new era of precision slide printing using laser technology.
  • Provides Improved barcode quality that enhances downstream workflow with improved scan ability
  • High resolution, 600dpi printing with LIS connectivity to streamline workflow
  • No additional consumables required such as ink, bulbs or ribbon
Reliable laser technology that can reduce instances of instrument downtime while delivering true on-demand slide barcode printing.
Certifications: Compliance with Laser Safety IEC 60825-1 and Electrical Safety IEC/EN 61010-1:2010.