2,3,4,5-Tetrahydro-1H-benzo[b]azepine 96%

Supplier: Ambeed
Warning

A165445-100MG A165445-250MG A165445-1G A165445-5G
103805-970EA 14 USD
103805-970 103844-224 77011-374 77011-376 77300-788 77320-434
2,3,4,5-Tetrahydro-1H-benzo[b]azepine 96%
2,3,4,5-Tetrahydro-1H-benzo[b]azepine

Caution: for Research Use Only

Formula: C₁₀H₁₃N
Boiling Pt: 268.9 °C (760 mm Hg)
Melting Pt: 30…34 °C
Flash Pt: 122.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00272363
CAS Number: 1701-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR