2-Amino-5-bromo-3-methylpyrazine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-3-methylpyrazin-2-amine
A163949-100MG
A163949-1G
A163949-5G
A163949-250MG
A163949-25G
103805-794EA
58
USD
103805-794
103844-060
76783-986
77320-182
77320-184
77667-292
77871-824
2-Amino-5-bromo-3-methylpyrazine 98%
2-Amino-5-bromo-3-methylpyrazine
Caution: for Research Use Only
Formula:
C₅H₆BrN₃ Boiling Pt: 272.4 °C (760 mm Hg) Flash Pt: 118.532 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08705773 CAS Number: 74290-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...