9-Bromoanthracene 98%

Supplier: Ambeed
Warning

A157372-100G A157372-10G A157372-500G A157372-25G A157372-5G
103843-198EA 5 USD
103843-198 76783-682 76783-684 76783-686 77300-254 77319-016
9-Bromoanthracene 98%
9-Bromoanthracene

Caution: for Research Use Only

Formula: C₁₄H₉Br
Boiling Pt: 389.7 °C (760 mm Hg)
Melting Pt: 97…100 °C
Flash Pt: 190.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00001243
CAS Number: 1564-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR