(2S,4S)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 1-tert-Butyl 2-Methyl (2S,4S)-4-Hydroxypyrrolidine-1,2-dicarboxylate, (2S,4S)-1-tert-Butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, (2S,4S)-4-Hydroxypyrrolidine-1,2-dicarboxylic Acid 1-tert-Butyl 2-Methyl Ester, N-Boc-cis-4-hydroxy-L-proline Methyl Ester, Boc-cis-Hyp-OMe

A156967-100G A156967-10G A156967-500G A156967-25G A156967-5G
103843-148EA 10 USD
103843-148 77028-438 77028-440 77028-442 77318-956 77318-958
(2S,4S)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate 97%
(2S,4S)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate

Caution: for Research Use Only

Formula: C₁₁H₁₉NO₅
Melting Pt: 82…86 °C
Flash Pt: 156.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00237541
CAS Number: 102195-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR