4-Bromopyridinium chloride 95%

Supplier: Ambeed
Warning

Synonyms: 4-Bromopyridine HCl, 4-Bromopyridine hydrochloride

A156741-100G A156741-10G A156741-500G A156741-25G
103843-120EA 5 USD
103843-120 77028-406 77028-408 77028-410 77318-918
4-Bromopyridinium chloride 95%
4-Bromopyridinium chloride

Caution: for Research Use Only

Formula: C₅H₅BrClN
Melting Pt: 270 °C
Flash Pt: 65.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00012828
CAS Number: 19524-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR