4-Hexylphenylboronic acid 97%

Supplier: Ambeed
Warning

A153365-5G
103804-498EA 24 USD
103804-498 103842-670 77318-326
4-Hexylphenylboronic acid 97%
4-Hexylphenylboronic acid

Caution: for Research Use Only

Formula: C₁₂H₁₉BO₂
Boiling Pt: 342.6 °C (760 mm Hg)
Flash Pt: 161 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039029
CAS Number: 105365-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR