D-(-)-Ribose 97%

Supplier: Ambeed
Warning

A151409-100G A151409-25G A151409-500G A151409-5G A151409-1000G
103808-758EA 39 USD
103808-758 76973-124 76973-126 76973-128 76973-130 77317-972
D-(-)-Ribose 97%
D(-)-Ribose

Caution: for Research Use Only

Formula: C₅H₁₀O₅
Boiling Pt: 375.4 °C (760 mm Hg)
Melting Pt: 95 °C
Flash Pt: 180.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00135453
CAS Number: 50-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR