tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate 97%

Supplier: Ambeed
Warning

A151105-10G A151105-250MG A151105-1G A151105-25G A151105-5G
103804-232EA 29 USD
103804-232 103842-378 77010-760 77010-762 77317-918 77317-920 77317-922
tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate 97%
tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate

Caution: for Research Use Only

Formula: C₁₁H₁₇N₃O₂S
Boiling Pt: 412.8 °C (760 mm Hg)
Flash Pt: 203.4 °C
Storage Temperature: Ambient
MDL Number: MFCD08448157
CAS Number: 365996-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR