1-Methylcyclopentanol 97%
Supplier: Ambeed
|
Danger
|
A144549-10G
A144549-1G
A144549-25G
A144549-5G
103803-420EA
5
USD
103803-420
103841-496
77235-140
77299-398
77316-748
77316-750
1-Methylcyclopentanol 97%
1-Methylcyclopentanol
Caution: for Research Use Only
Formula:
C₆H₁₂O Boiling Pt: 137.1 °C (760 mm Hg) Melting Pt: 36…37 °C Flash Pt: 41 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001364 CAS Number: 1462-03-9 UN: 1325 ADR: 4.1,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...