(S)-Methyl-2-aminopropanoate hydrochloride 97%

Supplier: Ambeed
Warning

A142845-100G A142845-10G A142845-500G A142845-25G A142845-1000G
103834-250EA 5 USD
103834-250 76972-682 76972-684 76972-686 77316-466 77881-114
(S)-Methyl-2-aminopropanoate hydrochloride 97%
(S)-Methyl-2-aminopropanoate hydrochloride

Caution: for Research Use Only

Formula: C₄H₉NO₂·HCl
Melting Pt: 109…111 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063663
CAS Number: 2491-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR