Mono(boc)(PEO)-3-diamine, boc-DOOA 97%

Supplier: Ambeed
Warning

Synonyms: N-(tert-Butoxycarbonyl)-2,2'-(ethylenedioxy)diethylamine, N-Boc-3,6-dioxa-1,8-octanediamine, tert-Butyl [2-[2-(2-Aminoethoxy)ethoxy]ethyl]carbamate, [2-[2-(2-Aminoethoxy)ethoxy]ethyl]carbamic acid tert-Butyl ester, N-Boc-2,2'-(ethylenedioxy)diethylamine, N-(tert-Butoxycarbonyl)-3,6-dioxa-1,8-octanediamine, tert-Butyl (2-(2-(2-aminoethoxy)ethoxy)ethyl)carbamate

A142623-100G A142623-10G A142623-250MG A142623-1G A142623-25G A142623-5G
103803-198EA 16 USD
103803-198 103834-228 76689-042 76689-044 76689-046 76689-048 77316-434 77316-436 77316-438
Mono(boc)(PEO)-3-diamine, boc-DOOA 97%
Mono(boc)(PEO)-3-diamine, boc-DOOA

Caution: for Research Use Only

Formula: C₁₁H₂₄N₂O₄
MW: 248.3 g/mol
Boiling Pt: 365.2 °C (760 mm Hg)
Flash Pt: 174.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03788155
CAS Number: 153086-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR