2-Indolinone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Oxindole, Oxindole, Indolin-2-one, 2,3-Dihydro-2-indolone
A136297-100G
A136297-10G
A136297-1G
A136297-500G
A136297-25G
A136297-5G
103833-256EA
6
USD
103833-256
76811-522
76811-524
76811-526
76811-528
77315-176
77315-178
2-Indolinone 98%
2-Indolinone
Caution: for Research Use Only
Formula:
C₈H₇NO MW: 133.15 g/mol Boiling Pt: 313 Melting Pt: 123…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005711 CAS Number: 59-48-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...