Z-D-Ser(tBu)-OH 98%

Supplier: Ambeed
Warning

Synonyms: Cbz-D-Ser(TBu)-OH, Z-D-Ser(TBu)-OH, Nα-Benzyloxycarbonyl-O-tert-butyl-D-serine

A135861-100G A135861-250MG A135861-1G A135861-25G A135861-5G
103802-228EA 8 USD
103802-228 103833-194 77109-522 77109-524 77315-096 77315-098 77315-100
Z-D-Ser(tBu)-OH 98%
Z-D-Ser(tBu)-OH

Caution: for Research Use Only

Formula: C₁₅H₂₁NO₅
MW: 295.335 g/mol
Boiling Pt: 465.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00077038
CAS Number: 65806-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR