2-Chloro-6-iodoaniline 98%
Supplier: Ambeed
|
Warning
|
A134005-10G
A134005-250MG
A134005-25G
A134005-1G
A134005-5G
103801-952EA
27
USD
103801-952
103832-892
77258-558
77258-560
77298-676
77314-660
77314-662
2-Chloro-6-iodoaniline 98%
2-Chloro-6-iodoaniline
Caution: for Research Use Only
Formula:
C₆H₅ClIN MW: 253.4681 g/mol Boiling Pt: 273.1 °C (760 mm Hg) Melting Pt: 66…67 °C Flash Pt: 119 °C Storage Temperature: Ambient |
MDL Number:
MFCD08443948 CAS Number: 84483-28-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...