4-Iodo-1H-pyrazole 97%

Supplier: Ambeed
Warning

A133653-100G A133653-10G A133653-500G A133653-1G A133653-25G A133653-1000G
103801-894EA 7 USD
103801-894 103832-828 76972-234 76972-236 76972-238 77298-646 77314-568 77670-906
4-Iodo-1H-pyrazole 97%
4-Iodo-1H-pyrazole

Caution: for Research Use Only

Formula: C₃H₃IN₂
Boiling Pt: 291.8 °C (760 mm Hg)
Melting Pt: 108…110 °C
Flash Pt: 130.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00005244
CAS Number: 3469-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR