4-Amino-2-fluorobenzoic acid 97%
Supplier: Ambeed
|
Danger
|
A133415-100G
A133415-1G
A133415-25G
A133415-5G
103801-874EA
6
USD
103801-874
103832-800
77136-976
77136-978
77314-520
77314-522
77314-524
4-Amino-2-fluorobenzoic acid 97%
4-Amino-2-fluorobenzoic acid
Caution: for Research Use Only
Formula:
C₇H₆FNO₂ MW: 155.13 Boiling Pt: 336.1 °C (760 mm Hg) Melting Pt: 210 °C Flash Pt: 157 °C Storage Temperature: Ambient |
MDL Number:
MFCD01569397 CAS Number: 446-31-1 UN: 1759 ADR: 8 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...