4-Amino-2-fluorobenzoic acid 97%

Supplier: Ambeed
Danger

A133415-100G A133415-1G A133415-25G A133415-5G
103801-874EA 6 USD
103801-874 103832-800 77136-976 77136-978 77314-520 77314-522 77314-524
4-Amino-2-fluorobenzoic acid 97%
4-Amino-2-fluorobenzoic acid

Caution: for Research Use Only

Formula: C₇H₆FNO₂
MW: 155.13
Boiling Pt: 336.1 °C (760 mm Hg)
Melting Pt: 210 °C
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD01569397
CAS Number: 446-31-1
UN: 1759
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR