Resiquimod 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-[4-Amino-2-(ethoxymethyl)-1H-imidazo[4,5-c]quinolin-1-yl]-2-methylpropan-2-ol, 4-Amino-2-(ethoxymethyl)-a,a-dimethyl-1H-imidazo[4,5-c]quinoline-1-ethanol, 1-(4-Amino-2-(ethoxymethyl)-1H-imidazo[4,5-c]quinolin-1-yl)-2-methylpropan-2-ol, 4-Amino-2-(ethoxymethyl)-alpha,alpha-dimethyl-1H-imidazo[4,5-c]quinoline-1-ethanol, R-848
A119633-100MG
A119633-1MG
A119633-250MG
A119633-5G
A119633-1G
103792-066EA
40
USD
103792-066
103792-068
103799-806
103799-306
76810-404
76810-406
76810-408
76810-410
77311-916
Resiquimod 98%
Resiquimod
Caution: for Research Use Only
Formula:
C₁₇H₂₂N₄O₂ Boiling Pt: 553.6 °C (760 mm Hg) Melting Pt: 166 °C Flash Pt: 288.6 °C Storage Temperature: Freezer |
MDL Number:
MFCD00937759 CAS Number: 144875-48-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...