Tris(dibenzylideneacetone)dipalladium(0) 98%

Supplier: Ambeed
Danger

Synonyms: Pd2(dba)3, Pd2dba3, Tris(dibenzylideneacetonyl)bis-palladium, Bis[tris(dibenzylideneacetone)palladium(0)]

A118340-10G A118340-250MG A118340-25G A118340-1G A118340-5G A118340-100G
103799-038EA 25 USD
103799-038 103830-406 76971-352 76971-354 76971-356 77311-716 77311-718 77566-166
Tris(dibenzylideneacetone)dipalladium(0) 98%
Tris(dibenzylideneacetone)dipalladium(0)

Caution: For research use only.

Formula: C₅₁H₄₂O₃Pd₂
Boiling Pt: 400.7 °C (760 mm Hg)
Melting Pt: 152…155 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013310
CAS Number: 51364-51-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR