3-Bromo-7-nitro-1H-indazole 98%
Supplier: Ambeed
|
Warning
|
A114732-100G
A114732-10G
A114732-250MG
A114732-25G
A114732-1G
A114732-5G
103798-460EA
11
USD
103798-460
103824-302
77271-158
77271-160
77271-162
77271-164
77311-122
77311-124
3-Bromo-7-nitro-1H-indazole 98%
3-Bromo-7-nitro-1H-indazole
Caution: for Research Use Only
Formula:
C₇H₄BrN₃O₂ Boiling Pt: 248.9 °C (760 mm Hg) Melting Pt: 175…185 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00159910 CAS Number: 74209-34-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...