Fmoc-L-Tyr(Me)-OH 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-Tyr(Me)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-methyl-L-tyrosine, N-Fmoc-4-methoxy-L-phenylalanine, N-Fmoc-O-methyl-L-tyrosine, Fmoc-Phe(4-OMe)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-methoxy-L-phenylalanine
A108968-250MG
A108968-1G
A108968-5G
A108968-10G
103797-418EA
6
USD
103797-418
103823-206
76869-622
77310-038
77310-040
77549-100
Fmoc-L-Tyr(Me)-OH 95%
Fmoc-L-Tyr(Me)-OH
Caution: for Research Use Only
Formula:
C₂₅H₂₃NO₅ MW: 417.47 g/mol Boiling Pt: 647.8 °C (760 mm Hg) Flash Pt: 345.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00153368 CAS Number: 77128-72-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...