Ethyl nicotinate 98%

Supplier: Ambeed
Warning

A107966-100G A107966-10G A107966-25G A107966-500G A107966-1000G
103807-986EA 51 USD
103807-986 77107-786 77107-788 77107-790 77107-792 77309-890
Ethyl nicotinate 98%
Ethyl nicotinate

Caution: for Research Use Only

Formula: C₈H₉NO₂
Boiling Pt: 224 °C (760 mm Hg)
Melting Pt: 8…10 °C
Flash Pt: 93.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00006389
CAS Number: 614-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR