1H-Pyrazole-1-carboximidamide hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 1-Amidinopyrazole Hydrochloride, 1H-Pyrazole-1-carboxamidine monohydrochloride, Hydrochloride, 1-Pyrazolecarboxamidine, Praxadine

A107913-100G A107913-10G A107913-25G A107913-500G A107913-5G
103807-980EA 337 USD
103807-980 103823-036 77233-290 77233-292 77233-294 77233-296 77309-882
1H-Pyrazole-1-carboximidamide hydrochloride 97%
1H-Pyrazole-1-carboximidamide hydrochloride

Caution: for Research Use Only

Formula: C₄H₇ClN₄
MW: 146.58 g/mol
Melting Pt: 7…170 °C
Flash Pt: 493 °C
Storage Temperature: Ambient
MDL Number: MFCD00210087
CAS Number: 4023-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR