Isonicotinaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: Pyridine-4-carboxaldehyde, 4-Pyridinecarboxaldehyde, 4-Pyridinecarbaldehyde

A105755-100G A105755-10G A105755-25G A105755-500G A105755-5G
103822-672EA 5 USD
103822-672 77107-570 77107-572 77107-574 77107-576 77309-538
Isonicotinaldehyde 98%
Isonicotinaldehyde

Caution: for Research Use Only

Formula: C₆H₅NO
Boiling Pt: 71…73 °C (10 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD00006425
CAS Number: 872-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR