SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate) 97%

Supplier: Ambeed
Warning

A104275-250MG A104275-1G A104275-5G A104275-100G A104275-25G
103796-678EA 17 USD
103796-678 103822-388 76717-704 77309-256 77309-258 77871-399 77871-400
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate) 97%
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)

Caution: for Research Use Only

Formula: C₁₆H₁₈N₂O₆
Boiling Pt: 501.7 °C (760 mm Hg)
Melting Pt: 180…182 °C
Flash Pt: 257.2 °C
Storage Temperature: Freezer
MDL Number: MFCD00009634
CAS Number: 64987-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR