Gamma Carbins Program for Nitrile Gloves
Supplier: POLYCARBIN, INC.
PR01-03-01
76716-896PK 621.46 USD
76716-896 77308-584
Gamma Carbins Program for Nitrile Gloves
Boxes Waste Disposal Boxes
Tired of nitrile gloves piling up in the trash? So are we. With Polycarbin's Gamma Carbin program, nitrile gloves will be used in the production of secondary goods, keeping them out of the landfill and incinerator.
  • For nitrile gloves only
  • Accepts all brands (and colors) of nitrile gloves
  • Access to personalized dashboard showing impact data
  • Prepaid return labels and carbon neutral shipping
  • Box designed to fit under lab bench
For every pound of nitrile gloves produced, the glove industry releases 5.5 pounds of CO₂ into the atmosphere. Polycarbin's Gamma Carbin program is a simple mailback service for non-infectious, non-hazardous nitrile gloves. Simply pack the Carbin with non-contaminated nitrile gloves. Once full, use the prepaid return labels to ship them back to us. From here, efforts can be tracked with Polycarbin's data portal, the Carbin Counter.
Delivery information: Gamma Carbins should come with prepaid return labels affixed to each box. If a Carbin does not have them upon arrival, please contact a Avantor representative.
Packaging: Gamma Carbins come in 5 packs and 2 packs. The external package dimensions are 27L×25W×10" H and 27L×25W×5" H, respectively.
Caution: The Gamma Carbin is exclusively for non-infectious, non-hazardous nitrile gloves. For a list of non-conforming items, please visit https://polycarbin.com/pages/gamma-carbin.