2,5-Dichloro-3-nitropyridine 98%

Supplier: Ambeed
Warning

A566151-25G A566151-100G A566151-10G A566151-1G
103828-774EA 5 USD
103828-774 103839-914 76844-028 77005-234 77005-236 77307-828
2,5-Dichloro-3-nitropyridine 98%
2,5-Dichloro-3-nitropyridine

Caution: for Research Use Only

Formula: C₅H₂Cl₂N₂O₂
MW: 192.99 g/mol
Boiling Pt: 265.3 °C (760 mm Hg)
Melting Pt: 39…45 °C
Flash Pt: 114.245 °C
Storage Temperature: Ambient
MDL Number: MFCD06658963
CAS Number: 21427-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR