Isophthalonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 1,3-Benzenedicarbonitrile, 1,3-Dicyanobenzene, Benzene-1,3-dicarbonitrile

A310400-100G A310400-500G A310400-25G A310400-1000G
76802-748EA 15 USD
76802-748 76802-750 77306-022 77668-372
Isophthalonitrile 98%
Isophthalonitrile

Caution: for Research Use Only

Formula: C₈H₄N₂
MDL Number: MFCD00001795
CAS Number: 626-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR