Ruthenium(III) chloride 97%

Supplier: Ambeed
Danger

Synonyms: Ruthenium trichloride

A275839-10G A275839-1G A275839-25G A275839-5G
103813-280EA 28 USD
103813-280 103853-206 76723-142 77305-214 77305-216 77305-218
Ruthenium(III) chloride 97%
Ruthenium (III) chloride

Caution: for Research Use Only

Formula: Cl₃Ru
Melting Pt: 500 °C
Storage Temperature: Ambient
MDL Number: MFCD00011208
CAS Number: 10049-08-8
UN: 3260
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR