Trithiocyanuric acid 98%

Supplier: Ambeed
Warning

A250986-100G A250986-10G A250986-500G A250986-25G
76786-822EA 34 USD
76786-822 76786-824 76786-826 77304-546
Trithiocyanuric acid 98%
Trithiocyanuric acid

Caution: for Research Use Only

Formula: C₃H₃N₃S₃
MW: 177.27 g/mol
MDL Number: MFCD00006052
CAS Number: 638-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR