DL-Glutamine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-DL-Gln-OH, 2,5-Diamino-5-oxopentanoic acid
A228319-100G
A228319-500G
A228319-25G
A228319-5G
103850-296EA
21
USD
103850-296
76987-226
76987-228
77303-828
77303-830
DL-Glutamine 97%
DL-Glutamine
Caution: for Research Use Only
Formula:
C₅H₁₀N₂O₃ Boiling Pt: 445.6 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD00065103 CAS Number: 6899-04-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...