9-Fluorenylmethanol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(Fluoren-9-yl)methanol, 9-Fluorenmethanol, 9-Fluorenemethanol
A221246-100G
A221246-500G
A221246-25G
103809-562EA
135
USD
103809-562
76999-920
76999-922
77303-586
9-Fluorenylmethanol 98%
9-Fluorenylmethanol
Caution: for Research Use Only
Formula:
C₁₄H₁₂O Boiling Pt: 363.9 °C (760 mm Hg) Melting Pt: 105…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001139 CAS Number: 24324-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...