4,7-Dihydroxy-1,10-phenanthroline 97%

Supplier: Ambeed
Warning

A220845-100MG A220845-10G A220845-250MG A220845-1G A220845-25G A220845-5G
103818-874EA 33 USD
103818-874 103849-756 77288-218 77288-220 77288-222 77303-560 77303-562 77303-564
4,7-Dihydroxy-1,10-phenanthroline 97%
4,7-Dihydroxy-1,10-phenanthroline

Caution: for Research Use Only

Formula: C₁₂H₈N₂O₂
Melting Pt: >300 °C
Flash Pt: 187.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00004975
CAS Number: 3922-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR