3-Cyano-4-methylpyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Methylpyridine-3-carbonitrile, 3-Cyano-4-picoline, 4-Methylnicotinonitrile
A205212-25G
A205212-5G
A205212-100G
A205212-10G
A205212-500G
A205212-1G
103817-618EA
7
USD
103817-618
103848-284
77086-490
77086-492
77274-978
77274-980
77274-982
77302-950
3-Cyano-4-methylpyridine 98%
3-Cyano-4-methylpyridine
Caution: for Research Use Only
Formula:
C₇H₆N₂ Boiling Pt: 231.7 °C (760 mm Hg) Melting Pt: 41…44 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234272 CAS Number: 5444-01-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...