Fmoc-D-Thr(tBu)-OH 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-D-threonine, Fmoc-D-Thr(tBu)-OH, (2R,3S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(tert-butoxy)butanoic acid, N-Fmoc-O-tert-butyl-D-threonine
A183224-10G
A183224-250MG
A183224-1G
A183224-25G
A183224-5G
A183224-100G
103815-620EA
7
USD
103815-620
103846-182
76784-658
76784-660
77301-824
77301-826
77301-828
77550-174
Fmoc-D-Thr(tBu)-OH 97%
Fmoc-D-Thr(tBu)-OH
Caution: for Research Use Only
Formula:
C₂₃H₂₇NO₅ MW: 397.471 g/mol Boiling Pt: 581.7 °C (760 mm Hg) Melting Pt: 130 °C Flash Pt: 305.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00080292 CAS Number: 138797-71-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...