1,3-Dibromoadamantane 97%
Supplier: Ambeed
|
Warning
|
A174152-1G
A174152-100G
A174152-10G
A174152-25G
A174152-5G
103806-922EA
6
USD
103806-922
103845-220
77082-508
77191-512
77191-514
77301-320
77301-322
1,3-Dibromoadamantane 97%
1,3-Dibromoadamantane
Caution: for Research Use Only
Formula:
C₁₀H₁₄Br₂ Boiling Pt: 287.6 °C (760 mm Hg) Melting Pt: 112…113 °C Flash Pt: 145.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077218 CAS Number: 876-53-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...