Tris(dibenzylideneacetone)dipalladium(0) 98%

Supplier: Ambeed
Warning

Synonyms: Pd2(dba)3, Pd2dba3, Tris(dibenzylideneacetonyl)bis-palladium, Bis[tris(dibenzylideneacetone)palladium(0)]

A172297-10G A172297-250MG A172297-25G A172297-5G A172297-1G
103806-710EA 47 USD
103806-710 103844-994 76647-232 76647-234 77082-184 77082-186 77301-192
Tris(dibenzylideneacetone)dipalladium(0) 98%
Tris(dibenzylideneacetone)dipalladium(0)

Caution: for Research Use Only

Formula: C₅₁H₄₂O₃Pd₂
Melting Pt: 152…155 °C
Flash Pt: 176.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013310
CAS Number: 51364-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR